ENA341294916 Enamine

5-(3-fluorophenyl)-1,3-oxazole-4-carboxylic acid

  • MDL number MFCD07376193
  • Cas Number : 1083401-22-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90630

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90630
Availability details for
Quantity
  • Availability
  • Add to Cart