ENA341294918 Enamine

3-(methanesulfonylmethyl)-1-benzothiophene-2-carboxylic acid

  • MDL number MFCD12858551
  • Cas Number : 1178235-42-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90632

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90632
Availability details for
Quantity
  • Availability
  • Add to Cart