ENA341294919 Enamine

trimethylpyrimidin-4-ol

  • MDL number MFCD07357440
  • Cas Number : 89943-15-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90633

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90633
Availability details for
Quantity
  • Availability
  • Add to Cart