ENA341294923 Enamine

1-(2-ethoxyethoxy)propan-2-ol

  • MDL number MFCD01696932
  • Cas Number : 104562-42-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90638

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90638
Availability details for
Quantity
  • Availability
  • Add to Cart