ENA341294927 Enamine

5-tert-butyl-1-(2-chloro-4-fluorophenyl)-1H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD13654587
  • Cas Number : 1097051-08-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90642

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90642
Availability details for
Quantity
  • Availability
  • Add to Cart