ENA341294928 Enamine

5-(2-chlorophenyl)-1,3-oxazole-4-carboxylic acid

  • MDL number MFCD01934491
  • Cas Number : 255876-53-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 194...196
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90643

Safety Information

msds https://www.enaminestore.com/catalog/EN300-90643
Availability details for
Quantity
  • Availability
  • Add to Cart