ENA341294931 Enamine

3-(trifluoromethanesulfonamido)propanoic acid

  • MDL number MFCD11649572
  • Cas Number : 141956-37-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90646

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90646
Availability details for
Quantity
  • Availability
  • Add to Cart