ENA341294932 Enamine

4,6-dichloro-5-ethyl-2-methylpyrimidine

  • MDL number MFCD11113325
  • Cas Number : 175139-08-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90659

Safety Information

msds https://www.enaminestore.com/catalog/EN300-90659
Availability details for
Quantity
  • Availability
  • Add to Cart