ENA341294936 Enamine

3-methyl-4-(trifluoromethyl)aniline

  • MDL number MFCD18399852
  • Cas Number : 106877-31-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91012

Safety Information

signalword Danger
pcodes P201,P202,P264,P270,P273,P280,P281,P301+P310,P305+P351+P338,P308+P313,P330,P337+P313,P391,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-91012
pictograms GHS06,GHS08,GHS09
hstatements H301,H319,H341,H400,H410
Availability details for
Quantity
  • Availability
  • Add to Cart