ENA341294944 Enamine

3,4-diphenylbutanoic acid

  • MDL number MFCD00094401
  • Cas Number : 4374-49-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 91-95
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91149

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91149
Availability details for
Quantity
  • Availability
  • Add to Cart