ENA341294947 Enamine

2-{2-[(4-cyanophenyl)methanesulfonamido]acetamido}acetic acid

  • MDL number MFCD13678384
  • Cas Number : 1156425-42-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91152

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91152
Availability details for
Quantity
  • Availability
  • Add to Cart