ENA341294949 Enamine

3-[(1,1-dioxo-1$l^{6},2-benzothiazol-3-yl)amino]butanoic acid

  • MDL number MFCD11550314
  • Cas Number : 1094565-75-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 245...247
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91156

Safety Information

msds https://www.enaminestore.com/catalog/EN300-91156
Availability details for
Quantity
  • Availability
  • Add to Cart