ENA341294950 Enamine

1-(4-methylphenyl)-1H-1,3-benzodiazol-2-amine hydrochloride

  • MDL number MFCD20441714
  • Cas Number : 1354963-37-3
  • Molecular Weight : 259.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-91158

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91158
Availability details for
Quantity
  • Availability
  • Add to Cart