ENA341294951 Enamine

2,2,2-trifluoroacetic acid; 3-amino-1-methylpyrrolidine-2,5-dione

  • MDL number MFCD20441715
  • Cas Number : 1354950-81-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 176...178
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91159

Safety Information

msds https://www.enaminestore.com/catalog/EN300-91159
Availability details for
Quantity
  • Availability
  • Add to Cart