ENA341294957 Enamine

methyl 2-(4-methoxyphenyl)propanoate

  • MDL number MFCD16748931
  • Cas Number : 50415-73-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-91171

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91171
Availability details for
Quantity
  • Availability
  • Add to Cart