ENA341294959 Enamine

2,2-dimethylbutanal

  • MDL number MFCD00055628
  • Cas Number : 2094-75-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91173

Safety Information

msds https://www.enaminestore.com/catalog/EN300-91173
Availability details for
Quantity
  • Availability
  • Add to Cart