ENA341294961 Enamine

1-bromo-3-(2,2-difluoroethoxy)benzene

  • MDL number MFCD11036117
  • Cas Number : 958454-25-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91175

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91175
Availability details for
Quantity
  • Availability
  • Add to Cart