ENA341294964 Enamine

2-(chloromethyl)-5-methyl-1,3-oxazole

  • MDL number MFCD03923820
  • Cas Number : 731851-14-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91180

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91180
Availability details for
Quantity
  • Availability
  • Add to Cart