ENA341294972 Enamine

1-tert-butyl-3-(2-ethoxyphenyl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD13654235
  • Cas Number : 1152891-41-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 207...209
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91192

Safety Information

msds https://www.enaminestore.com/catalog/EN300-91192
Availability details for
Quantity
  • Availability
  • Add to Cart