ENA341294984 Enamine

2-(1H-1,2,3,4-tetrazol-5-yl)propan-2-amine

  • MDL number MFCD12796286
  • Cas Number : 1211587-69-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 251...253
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91243

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-91243
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart