ENA341294993 Enamine

4-[(4-methylcyclohexyl)amino]cyclohexan-1-ol

  • MDL number MFCD06620006
  • Cas Number : 1152964-08-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 143...145
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91254

Safety Information

msds https://www.enaminestore.com/catalog/EN300-91254
Availability details for
Quantity
  • Availability
  • Add to Cart