ENA341294995 Enamine

1-[3-(trifluoromethyl)phenyl]-1H-pyrazole-4-sulfonyl chloride

  • MDL number MFCD12478347
  • Cas Number : 1156915-96-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91257

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91257
Availability details for
Quantity
  • Availability
  • Add to Cart