ENA341294996 Enamine

1-(2,3-dihydro-1,4-benzodioxin-6-yl)-1H-pyrrole-2-carboxylic acid

  • MDL number MFCD12072561
  • Cas Number : 1155519-16-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91260

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91260
Availability details for
Quantity
  • Availability
  • Add to Cart