ENA341294999 Enamine

5-methyl-2-(2-methylphenyl)morpholine

  • MDL number MFCD11653033
  • Cas Number : 1099679-79-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91274

Safety Information

msds https://www.enaminestore.com/catalog/EN300-91274
Availability details for
Quantity
  • Availability
  • Add to Cart