ENA341295008 Enamine

3-(trifluoromethyl)oxolan-3-ol

  • MDL number MFCD18333905
  • Cas Number : 1269151-92-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90126

Safety Information

msds https://www.enaminestore.com/catalog/EN300-90126
Availability details for
Quantity
  • Availability
  • Add to Cart