ENA341295016 Enamine

methyl 3-(2H-1,2,3,4-tetrazol-5-yl)benzoate

  • MDL number MFCD09263263
  • Cas Number : 148345-63-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 189...191
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91317

Safety Information

msds https://www.enaminestore.com/catalog/EN300-91317
Availability details for
Quantity
  • Availability
  • Add to Cart