ENA341295036 Enamine

[4-(3-methoxyphenyl)phenyl]methanamine

  • MDL number MFCD08701379
  • Cas Number : 858674-00-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 46...48
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91351

Safety Information

msds https://www.enaminestore.com/catalog/EN300-91351
Availability details for
Quantity
  • Availability
  • Add to Cart