ENA341295038 Enamine

2-(3-amino-1H-1,2,4-triazol-1-yl)-N-methylacetamide

  • MDL number MFCD12804840
  • Cas Number : 1182757-96-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149...151
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91354

Safety Information

msds https://www.enaminestore.com/catalog/EN300-91354
Availability details for
Quantity
  • Availability
  • Add to Cart