ENA341295052 Enamine

4-methyl-N-[(oxolan-2-yl)methyl]cyclohexan-1-amine

  • MDL number MFCD01453750
  • Cas Number : 467444-21-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91369

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91369
Availability details for
Quantity
  • Availability
  • Add to Cart