ENA341295054 Enamine

3-oxo-3-(oxolan-3-yl)propanenitrile

  • MDL number MFCD14610497
  • Cas Number : 1186610-03-6
  • Molecular Weight : 139.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form oil
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91371

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P301+P310,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-91371
pictograms GHS06
hstatements H301,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart