ENA341295061 Enamine

1-(aminomethyl)-5,6,7,8-tetrahydronaphthalen-2-ol hydrochloride

  • MDL number MFCD20441730
  • Cas Number : 1354952-20-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91409

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91409
Availability details for
Quantity
  • Availability
  • Add to Cart