ENA341295074 Enamine

2-chloro-6-(4-fluorophenyl)pyrazine

  • MDL number MFCD20441735
  • Cas Number : 1273550-42-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 95...97
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91425

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-91425
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart