ENA341295079 Enamine

1-(2-fluorophenyl)-1H-1,3-benzodiazol-2-amine

  • MDL number MFCD11202310
  • Cas Number : 1038303-09-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91430

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91430
Availability details for
Quantity
  • Availability
  • Add to Cart