ENA341295088 Enamine

2-amino-6-bromo-4-methylphenol

  • MDL number MFCD09754409
  • Cas Number : 343269-51-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91440

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-91440
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart