ENA341295090 Enamine

2-(3-fluorophenyl)-2-(morpholin-4-yl)acetic acid hydrochloride

  • MDL number MFCD20441740
  • Cas Number : 1354950-40-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91443

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91443
Availability details for
Quantity
  • Availability
  • Add to Cart