ENA341295107 Enamine

3-chloro-4-(chloromethyl)pyridine hydrochloride

  • MDL number MFCD20441743
  • Cas Number : 132685-21-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91493

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91493
Availability details for
Quantity
  • Availability
  • Add to Cart