ENA341295111 Enamine

5-chloro-1-(2-fluorophenyl)-3-propyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD12739384
  • Cas Number : 1152978-92-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91500

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91500
Availability details for
Quantity
  • Availability
  • Add to Cart