ENA341295135 Enamine

2,3,4,5,6-pentafluorophenyl 4,5-dimethyl-1,3-thiazole-2-sulfonate

  • MDL number MFCD20441747
  • Cas Number : 1354950-61-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91532

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91532
Availability details for
Quantity
  • Availability
  • Add to Cart