ENA341295139 Enamine

5-{4H-imidazo[4,5-b]pyridin-2-yl}quinoline

  • MDL number MFCD26143901
  • Cas Number : 1354960-76-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91536

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91536
Availability details for
Quantity
  • Availability
  • Add to Cart