ENA341295152 Enamine

4-(prop-2-en-1-yloxy)phenol

  • MDL number MFCD02113809
  • Cas Number : 6411-34-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-91576

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91576
Availability details for
Quantity
  • Availability
  • Add to Cart