ENA341295157 Enamine

4-(N-methylacetamido)benzene-1-sulfonyl chloride

  • MDL number MFCD10037346
  • Cas Number : 39169-92-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91596

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91596
Availability details for
Quantity
  • Availability
  • Add to Cart