ENA341295165 Enamine

1-(3-chlorophenyl)-2-oxopyrrolidine-3-carboxylic acid

  • MDL number MFCD03932880
  • Cas Number : 618070-35-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 113...115
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91607

Safety Information

msds https://www.enaminestore.com/catalog/EN300-91607
Availability details for
Quantity
  • Availability
  • Add to Cart