ENA341295174 Enamine

1,4-diazaspiro[5.5]undecane

  • MDL number MFCD07373058
  • Cas Number : 180-76-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91624

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91624
Availability details for
Quantity
  • Availability
  • Add to Cart