ENA341295182 Enamine

1-[(thiophen-3-yl)methyl]piperidin-4-ol

  • MDL number MFCD01652491
  • Cas Number : 864388-84-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91632

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91632
Availability details for
Quantity
  • Availability
  • Add to Cart