ENA341295183 Enamine

2-phenyl-2-(trifluoromethyl)pyrrolidine

  • MDL number MFCD18380773
  • Cas Number : 1269151-41-8
  • Molecular Weight : 215.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-90135

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90135
Availability details for
Quantity
  • Availability
  • Add to Cart