ENA341295190 Enamine

1-(5-ethyl-1,3-thiazol-2-yl)piperazine

  • MDL number MFCD16834332
  • Cas Number : 1211541-43-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91642

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91642
Availability details for
Quantity
  • Availability
  • Add to Cart