ENA341295192 Enamine

3-(2,2,2-trifluoroethyl)pyrrolidine hydrochloride

  • MDL number MFCD18380776
  • Cas Number : 1269152-60-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90138

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90138
Availability details for
Quantity
  • Availability
  • Add to Cart