ENA341295195 Enamine

4-ethylpiperidin-4-ol hydrochloride

  • MDL number MFCD16875508
  • Cas Number : 1354949-87-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149...151
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91654

Safety Information

msds https://www.enaminestore.com/catalog/EN300-91654
Availability details for
Quantity
  • Availability
  • Add to Cart