ENA341295199 Enamine

methyl[1-(4-methylphenyl)butan-2-yl]amine

  • MDL number MFCD12154524
  • Cas Number : 861199-21-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91658

Safety Information

msds https://www.enaminestore.com/catalog/EN300-91658
Availability details for
Quantity
  • Availability
  • Add to Cart