ENA341295201 Enamine

[1-cyclopropyl-2-(4-methylphenyl)ethyl](ethyl)amine

  • MDL number MFCD12154665
  • Cas Number : 1178738-58-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91660

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91660
Availability details for
Quantity
  • Availability
  • Add to Cart